Assay Details
Target Gene Details
Entrez Gene ID: | 55529 |
Gene Name: | transmembrane protein 55A |
Gene Aliases: |
- |
Location: |
Chr.8:90994271-91040975 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM55A | NM_018710.2 | 7 | 935 | NP_061180.1 |
| AK313783.1 | 7 | 848 | ||
| AX775961.1 | 7 | 785 | CAE11680.1 | |
| BC033892.1 | 7 | 785 | AAH33892.1 | |
| CR749733.1 | 6 | 499 | CAH18492.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1028201 | Chr.8:90956299 - 90997736 on Build GRCh38 | Loss |
|
| esv2759629 | Chr.8:90962271 - 91261496 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map