Assay Details
Target Gene Details
Entrez Gene ID: | 54826 |
Gene Name: | gypsy retrotransposon integrase 1 |
Gene Aliases: |
GIN-1, TGIN1, ZH2C2 |
Location: |
Chr.5:103086000-103120149 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GIN1 | NM_001317954.1 | 2 | 163 | NP_001304883.1 |
| NM_017676.2 | 2 | 163 | NP_060146.2 | |
| NR_134299.1 | 2 | 163 | ||
| XM_005272028.3 | 2 | 174 | XP_005272085.1 | |
| XM_017009599.1 | 2 | 1598 | XP_016865088.1 | |
| AK000132.1 | 2 | 161 | BAA90963.1 | |
| AK225572.1 | 2 | 160 | ||
| AK295796.1 | 2 | 151 | ||
| BC015325.1 | 2 | 122 | AAH15325.1 | |
| BC151135.1 | 1 | 103 | ||
| BC157835.1 | 1 | 103 | ||
| CR627424.1 | 2 | 134 | CAH10511.1 | |
| DB101499.1 | 2 | 163 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830430 | Chr.5:102996203 - 103156228 on Build GRCh38 | Gain |
|
| nsv1029894 | Chr.5:102284375 - 103249595 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map