Assay Details
Target Gene Details
Entrez Gene ID: | 9506 |
Gene Name: | PAGE family member 4 |
Gene Aliases: |
CT16.7, GAGE-9, GAGEC1, JM-27, JM27, PAGE-1, PAGE-4 |
Location: |
Chr.X:49829260-49834264 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAGE4 | NM_001318877.1 | NP_001305806.1 | ||
| NM_007003.4 | NP_008934.1 | |||
| AF275258.1 | AAF88037.1 | |||
| AI093155.1 | ||||
| AJ005894.1 | CAA06751.1 | |||
| AK092503.1 | ||||
| AK312039.1 | ||||
| AL548759.3 | ||||
| BC010897.2 | AAH10897.1 | |||
| BG435968.1 | ||||
| CR456823.1 | CAG33104.1 | |||
| CR542183.1 | CAG46980.1 | |||
| DA841276.1 |
More Information
Set Membership: |
|

Genomic Map