Assay Details
Target Gene Details
Entrez Gene ID: | 11194 |
Gene Name: | ATP binding cassette subfamily B member 8 |
Gene Aliases: |
EST328128, M-ABC1, MABC1 |
Location: |
Chr.7:151028422-151047782 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCB8 | NM_001282291.1 | NP_001269220.1 | ||
| NM_001282292.1 | NP_001269221.1 | |||
| NM_001282293.1 | NP_001269222.1 | |||
| NM_007188.4 | NP_009119.2 | |||
| AF047690.1 | AAD15748.1 | |||
| AK002018.1 | BAA92038.1 | |||
| AK024401.1 | ||||
| AK094005.1 | 2 | 360 | ||
| AK094636.1 | BAC04392.1 | |||
| AK222911.1 | BAD96631.1 | |||
| AK294344.1 | ||||
| AK310796.1 | ||||
| AK314721.1 | ||||
| BC141814.1 | ||||
| BC141836.1 | ||||
| BC151235.1 | ||||
| U66688.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526410 | Chr.7:151017725 - 151078219 on Build GRCh38 | Loss |
|
| nsv951648 | Chr.7:150931413 - 151106413 on Build GRCh38 | Deletion |
|
| dgv6756n100 | Chr.7:151033912 - 151093430 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map