Assay Details
Target Gene Details
Entrez Gene ID: | 1017 |
Gene Name: | cyclin dependent kinase 2 |
Gene Aliases: |
CDKN2, p33(CDK2) |
Location: |
Chr.12:55966769-55972789 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDK2 | NM_001290230.1 | 5 | 1790 | NP_001277159.1 |
| NM_001798.4 | 7 | 1970 | NP_001789.2 | |
| NM_052827.3 | 6 | 1868 | NP_439892.2 | |
| XM_011537732.1 | 8 | 2114 | XP_011536034.1 | |
| AA789250.1 | 1 | 334 | ||
| BC003065.2 | 7 | 1890 | AAH03065.1 | |
| M68520.1 | 7 | 1920 | AAA35667.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832424 | Chr.12:55881442 - 56073133 on Build GRCh38 | Loss |
|
| nsv1051961 | Chr.12:55868436 - 56235643 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map