Assay Details
Target Gene Details
Entrez Gene ID: | 79982 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member B14 |
Gene Aliases: |
EGNR9427, PRO34683 |
Location: |
Chr.4:99896248-99946726 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJB14 | NM_001031723.3 | 8 | 2834 | NP_001026893.1 |
| NM_001278310.1 | 7 | 2662 | NP_001265239.1 | |
| XM_011532262.2 | 10 | 2718 | XP_011530564.1 | |
| XM_011532263.2 | 8 | 3218 | XP_011530565.1 | |
| XM_011532265.2 | 7 | 3162 | XP_011530567.1 | |
| XM_017008628.1 | 10 | 2900 | XP_016864117.1 | |
| AL833076.2 | 7 | 4706 | CAD89928.1 | |
| AL833412.1 | 7 | 2524 | ||
| AY186740.1 | 10 | 2713 | AAO31693.1 | |
| AY358272.1 | 8 | 2688 | AAQ88639.1 |

Genomic Map