Assay Details
Target Gene Details
Entrez Gene ID: | 79004 |
Gene Name: | CUE domain containing 2 |
Gene Aliases: |
C10orf66, bA18I14.5 |
Location: |
Chr.10:102423245-102432666 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CUEDC2 | NM_024040.2 | NP_076945.2 | ||
| AB025425.1 | BAB87809.1 | |||
| AB232358.1 | BAE19942.1 | |||
| BC000262.1 | AAH00262.1 | |||
| BF002772.1 | ||||
| CX165866.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1159761 | Chr.10:102372778 - 102430809 on Build GRCh38 | Deletion |
|
| nsv951006 | Chr.10:102394644 - 102425743 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map