Assay Details
Target Gene Details
Entrez Gene ID: | 1389 |
Gene Name: | cAMP responsive element binding protein like 2 |
Gene Aliases: |
- |
Location: |
Chr.12:12611833-12645108 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CREBL2 | NM_001310.3 | 1 | 215 | NP_001301.1 |
| AF039081.1 | 1 | 151 | AAC32314.1 | |
| AK307202.1 | 1 | 150 | ||
| BC104873.1 | 1 | 59 | AAI04874.1 | |
| BC106052.1 | 1 | 156 | AAI06053.1 | |
| BC112157.1 | 1 | 59 | AAI12158.1 | |
| BY795169.2 | 1 | 215 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759880 | Chr.12:12467720 - 12630190 on Build GRCh38 | Gain |
|
| esv2745567 | Chr.12:12246086 - 13092998 on Build GRCh38 | Deletion |
|
| nsv1046355 | Chr.12:11501836 - 12890185 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map