Assay Details
Target Gene Details
Entrez Gene ID: | 57821 |
Gene Name: | coiled-coil domain containing 181 |
Gene Aliases: |
C1orf114 |
Location: |
Chr.1:169394870-169462221 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC181 | NM_001300968.1 | 4 | 924 | NP_001287897.1 |
| NM_001300969.1 | 3 | 626 | NP_001287898.1 | |
| NM_021179.2 | 3 | 626 | NP_067002.1 | |
| XM_005245383.1 | 4 | 924 | XP_005245440.1 | |
| XM_011509827.2 | 3 | 608 | XP_011508129.1 | |
| XM_017001938.1 | 5 | 2557 | XP_016857427.1 | |
| XM_017001939.1 | 3 | 589 | XP_016857428.1 | |
| XM_017001940.1 | 3 | 608 | XP_016857429.1 | |
| AK098712.1 | 3 | 569 | ||
| AK223354.1 | 3 | 569 | BAD97074.1 | |
| AK313598.1 | 4 | 924 | ||
| AL049687.1 | 3 | 584 | CAB41258.1 | |
| BC026073.2 | 3 | 626 | AAH26073.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2663262 | Chr.1:169421907 - 169426804 on Build GRCh38 | Deletion |
|
| nsv1001454 | Chr.1:169418143 - 169496036 on Build GRCh38 | Gain |
|
| esv22143 | Chr.1:169090390 - 169652030 on Build GRCh38 | Gain+Loss |
|
| nsv831871 | Chr.1:169351435 - 169517442 on Build GRCh38 | Loss |
|

Genomic Map