Assay Details
Target Gene Details
Entrez Gene ID: | 10929 |
Gene Name: | serine and arginine rich splicing factor 8 |
Gene Aliases: |
DSM-1, SFRS2B, SRP46 |
Location: |
Chr.11:95066877-95071227 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRSF8 | NM_032102.3 | 1 | 1445 | NP_115285.1 |
| NR_103726.1 | ||||
| XM_017017142.1 | 1 | 1407 | XP_016872631.1 | |
| AF031166.1 | 1 | 1065 | AAK54351.1 | |
| AK023379.1 | ||||
| AK309776.1 | 3 | 757 | ||
| BC057783.1 | 2 | 1135 | AAH57783.1 | |
| CB215027.1 | 1 | 406 | ||
| CV805885.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs111408022] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map