Assay Details
Target Gene Details
Entrez Gene ID: | 128387 |
Gene Name: | TatD DNase domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.1:212791828-212816825 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TATDN3 | NM_001042552.2 | 6 | 468 | NP_001036017.1 |
| NM_001042553.2 | 6 | 468 | NP_001036018.1 | |
| NM_001146169.1 | 6 | 468 | NP_001139641.1 | |
| NM_001146170.1 | 5 | 405 | NP_001139642.1 | |
| NM_001146171.1 | 6 | 468 | NP_001139643.1 | |
| XM_011509202.2 | 6 | 463 | XP_011507504.1 | |
| XM_017000328.1 | 5 | 400 | XP_016855817.1 | |
| XM_017000329.1 | 5 | 400 | XP_016855818.1 | |
| XM_017000330.1 | 6 | 463 | XP_016855819.1 | |
| XM_017000331.1 | 6 | 463 | XP_016855820.1 | |
| AK293129.1 | 6 | 468 | ||
| AK295335.1 | 5 | 393 | ||
| AK304557.1 | 5 | 363 | ||
| AL832248.1 | 6 | 410 | ||
| BC048115.1 | 6 | 379 | AAH48115.1 | |
| BC113638.1 | 6 | 412 | ||
| BC117485.1 | 6 | 412 | ||
| BC143956.1 | 6 | 412 | ||
| DB456872.1 | 5 | 361 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758997 | Chr.1:212752087 - 212833055 on Build GRCh38 | Loss |
|
| nsv1010003 | Chr.1:212770898 - 212813654 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map