Assay Details
Target Gene Details
Entrez Gene ID: | 5876 |
Gene Name: | Rab geranylgeranyltransferase beta subunit |
Gene Aliases: |
GGTB |
Location: |
Chr.1:75786194-75795090 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RABGGTB | NM_004582.3 | NP_004573.2 | ||
| NR_073562.1 | ||||
| AB209023.1 | BAD92260.1 | |||
| BC020790.1 | AAH20790.1 | |||
| BQ773824.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519329 | Chr.1:75364753 - 75892906 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74966573] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map