Assay Details
Target Gene Details
Entrez Gene ID: | 57713 |
Gene Name: | Scm-like with four mbt domains 2 |
Gene Aliases: |
- |
Location: |
Chr.10:7158624-7411486 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SFMBT2 | NM_001018039.1 | 21 | 3833 | NP_001018049.1 |
| NM_001029880.2 | 21 | 3935 | NP_001025051.1 | |
| XM_005252551.2 | 21 | 3716 | XP_005252608.1 | |
| XM_006717490.2 | 19 | 3708 | XP_006717553.1 | |
| XM_011519611.1 | 21 | 3934 | XP_011517913.1 | |
| XM_017016466.1 | 21 | 3836 | XP_016871955.1 | |
| AB046837.1 | 21 | 3833 | ||
| BC152430.1 | 21 | 3833 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831777 | Chr.10:7133245 - 7298120 on Build GRCh38 | Gain |
|
| nsv1041863 | Chr.10:7063546 - 7188862 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map