Assay Details
Target Gene Details
Entrez Gene ID: | 9020 |
Gene Name: | mitogen-activated protein kinase kinase kinase 14 |
Gene Aliases: |
FTDCR1B, HS, HSNIK, NIK |
Location: |
Chr.17:45263119-45317064 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP3K14 | NM_003954.4 | NP_003945.2 | ||
| XM_011525441.2 | XP_011523743.1 | |||
| AJ008160.1 | ||||
| AJ008161.1 | ||||
| AK131438.1 | BAD18584.1 | |||
| AK290203.1 | ||||
| AK313085.1 | ||||
| BC035576.1 | AAH35576.1 | |||
| CR749592.1 | CAH18391.1 | |||
| Y10256.1 | CAA71306.1 |
Target Gene Details
Entrez Gene ID: | 100133991 |
Gene Name: | MAP3K14 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:45247925-45268630 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP3K14-AS1 | NR_024434.2 | |||
| NR_024435.2 | ||||
| NR_110324.1 | ||||
| NR_110325.1 | ||||
| NR_110326.1 | ||||
| AK097219.1 | ||||
| AK311429.1 | ||||
| AK311564.1 | ||||
| AL710937.1 | ||||
| DB133288.1 | ||||
| DB294632.1 | ||||
| DB466340.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953904 | Chr.17:45231034 - 45277033 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map