Assay Details
Target Gene Details
Entrez Gene ID: | 282809 |
Gene Name: | POC1 centriolar protein B |
Gene Aliases: |
CORD20, PIX1, TUWD12, WDR51B |
Location: |
Chr.12:89419718-89526262 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POC1B | NM_001199777.1 | 11 | 2696 | NP_001186706.1 |
| NM_172240.2 | 12 | 2715 | NP_758440.1 | |
| NR_037659.1 | 12 | 2566 | ||
| NR_037660.1 | 10 | 2524 | ||
| AB053266.1 | 11 | 2205 | BAB69057.1 | |
| AK027829.1 | 8 | 1914 | ||
| AK074772.1 | 12 | 2453 | BAC11198.1 | |
| AK123625.1 | 8 | 1913 | BAC85666.1 | |
| AL832918.1 | 11 | 2202 | CAH10775.1 | |
| BX648293.1 | 2 | 5317 |
More Information
Set Membership: |
|

Genomic Map