Assay Details
Target Gene Details
Entrez Gene ID: | 3141 |
Gene Name: | holocarboxylase synthetase |
Gene Aliases: |
HCS |
Location: |
Chr.21:36750888-36990254 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HLCS | NM_000411.6 | 12 | 5949 | NP_000402.3 |
| NM_001242784.1 | 12 | 6883 | NP_001229713.1 | |
| NM_001242785.1 | 12 | 6042 | NP_001229714.1 | |
| BI094308.1 | 1 | 502 | ||
| BQ775124.1 | 1 | 85 |
More Information
Set Membership: |
|

Genomic Map