Assay Details
Target Gene Details
Entrez Gene ID: | 56892 |
Gene Name: | chromosome 8 open reading frame 4 |
Gene Aliases: |
TC-1, TC1 |
Location: |
Chr.8:40153468-40155308 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C8orf4 | NM_020130.4 | 1 | 1036 | NP_064515.1 |
| AF268037.1 | 1 | 1020 | AAF78961.1 | |
| AK026240.1 | 1 | 1028 | ||
| BC020623.1 | 1 | 1022 | AAH20623.1 | |
| BC021672.1 | 1 | 1040 | AAH21672.1 | |
| BM976768.1 | 1 | 327 | ||
| BX434108.2 | 1 | 281 |
More Information
Set Membership: |
|

Genomic Map