Assay Details
Target Gene Details
Entrez Gene ID: | 55732 |
Gene Name: | chromosome 1 open reading frame 112 |
Gene Aliases: |
- |
Location: |
Chr.1:169794730-169854080 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1orf112 | NM_001320047.1 | 9 | 1027 | NP_001306976.1 |
| NM_001320048.1 | 7 | 782 | NP_001306977.1 | |
| NM_018186.3 | 8 | 1330 | NP_060656.2 | |
| XM_005245317.4 | 8 | 902 | XP_005245374.1 | |
| XM_011509735.2 | XP_011508037.1 | |||
| XM_017001722.1 | 8 | 902 | XP_016857211.1 | |
| XM_017001723.1 | 8 | 902 | XP_016857212.1 | |
| XM_017001724.1 | 6 | 651 | XP_016857213.1 | |
| XM_017001725.1 | XP_016857214.1 | |||
| AK001568.1 | 7 | 702 | BAA91761.1 | |
| AK096493.1 | 9 | 937 | ||
| AK127098.1 | 12 | 1724 | ||
| AK294569.1 | 4 | 473 | ||
| AK299369.1 | 8 | 813 | ||
| AK303296.1 | 7 | 778 | ||
| AK310638.1 | ||||
| AL354614.1 | 8 | 1330 | CAB89727.1 | |
| BC091516.1 | 9 | 941 | AAH91516.1 | |
| BC107168.1 | AAI07169.1 | |||
| BC107169.1 | 8 | 818 | AAI07170.1 |

Genomic Map