Assay Details
Target Gene Details
Entrez Gene ID: | 79637 |
Gene Name: | armadillo repeat containing 7 |
Gene Aliases: |
- |
Location: |
Chr.17:75109952-75130265 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARMC7 | NM_001304271.1 | 3 | 1761 | NP_001291200.1 |
| NM_024585.3 | 3 | 1825 | NP_078861.1 | |
| AK025813.1 | 3 | 1633 | BAB15242.1 | |
| BC011728.1 | 3 | 1790 | AAH11728.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470602 | Chr.17:75090871 - 75208042 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map