Assay Details
Target Gene Details
Entrez Gene ID: | 259282 |
Gene Name: | biorientation of chromosomes in cell division 1 like 1 |
Gene Aliases: |
BOD1L, FAM44A |
Location: |
Chr.4:13568738-13627721 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BOD1L1 | NM_148894.2 | 26 | 9988 | NP_683692.2 |
| XM_005248150.3 | 27 | 10146 | XP_005248207.1 | |
| XM_005248151.3 | 27 | 10143 | XP_005248208.1 | |
| XM_006713958.3 | 28 | 10188 | XP_006714021.1 | |
| XM_011513827.2 | 28 | 10191 | XP_011512129.1 | |
| XM_011513829.2 | 27 | 10050 | XP_011512131.1 | |
| XM_011513830.2 | 26 | 9574 | XP_011512132.1 | |
| XM_017008009.1 | 27 | 10047 | XP_016863498.1 | |
| XM_017008010.1 | 26 | 10002 | XP_016863499.1 | |
| AB037748.2 | 17 | 6132 | BAA92565.2 | |
| AK025361.1 | 12 | 1649 | ||
| AL137652.1 | 1 | 2593 | ||
| BC016987.1 | 16 | 1928 | AAH16987.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4239 | Chr.4:13564336 - 13622398 on Build GRCh38 | Insertion |
|
| nsv593750 | Chr.4:13259081 - 13726709 on Build GRCh38 | Gain |
|
| nsv1014473 | Chr.4:11586127 - 13990352 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map