Assay Details
Target Gene Details
Entrez Gene ID: | 164091 |
Gene Name: | progestin and adipoQ receptor family member 7 |
Gene Aliases: |
MPRA, PGLP, mSR |
Location: |
Chr.1:25861484-25876707 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAQR7 | NM_178422.5 | 2 | 1215 | NP_848509.1 |
| XM_005245745.3 | 3 | 1037 | XP_005245802.1 | |
| XM_005245746.3 | 2 | 953 | XP_005245803.1 | |
| XM_011540861.1 | 3 | 860 | XP_011539163.1 | |
| XM_011540862.2 | 4 | 1126 | XP_011539164.1 | |
| AF313620.1 | 2 | 644 | AAO47233.1 | |
| AY424285.1 | 1 | 776 | AAR08373.1 | |
| BC034015.1 | 2 | 1215 | AAH34015.1 | |
| BC127800.1 | 1 | 326 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv460884 | Chr.1:25778482 - 25891593 on Build GRCh38 | Loss |
|
| nsv834491 | Chr.1:25726411 - 25914117 on Build GRCh38 | Loss |
|
| nsv834502 | Chr.1:25834281 - 25987428 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map