Assay Details
Target Gene Details
Entrez Gene ID: | 998 |
Gene Name: | cell division cycle 42 |
Gene Aliases: |
CDC42Hs, G25K, TKS |
Location: |
Chr.1:22052627-22092943 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC42 | NM_001039802.1 | NP_001034891.1 | ||
| NM_001791.3 | NP_001782.1 | |||
| NM_044472.2 | NP_426359.1 | |||
| AF498962.1 | AAM21109.1 | |||
| AF498963.1 | AAM21110.1 | |||
| AK297465.1 | ||||
| AK303994.1 | ||||
| AK311375.1 | ||||
| AK316449.1 | ||||
| AL121734.1 | CAB57325.1 | |||
| AL121735.1 | CAB57326.1 | |||
| AL121736.1 | CAB57327.1 | |||
| AL121737.1 | CAB57328.1 | |||
| BC002711.2 | AAH02711.1 | |||
| BC003682.1 | AAH03682.1 | |||
| BC018266.1 | AAH18266.1 | |||
| BP871988.1 | ||||
| M35543.1 | AAA52494.1 | |||
| M57298.1 | AAA52592.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv954904 | Chr.1:22053808 - 22107507 on Build GRCh38 | Duplication |
|
| nsv945817 | Chr.1:22078110 - 22078778 on Build GRCh38 | Duplication |
|
| esv2758927 | Chr.1:21896028 - 22086364 on Build GRCh38 | Loss |
|
| nsv834413 | Chr.1:21908720 - 22093681 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map