Assay Details
Target Gene Details
Entrez Gene ID: | 6840 |
Gene Name: | supervillin |
Gene Aliases: |
- |
Location: |
Chr.10:29457338-29737001 on Build GRCh38 |
Assay Gene Location: | Within Exon 47 |
Target Gene Details
Entrez Gene ID: | 102724316 |
Gene Name: | SVIL antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:29409534-29487856 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SVIL-AS1 | NR_110920.1 | |||
| NR_110921.1 | ||||
| NR_110922.1 | ||||
| NR_110925.1 | ||||
| NR_110926.1 | ||||
| NR_110927.1 | ||||
| NR_110928.1 | ||||
| NR_110929.1 | ||||
| BF511092.1 | ||||
| BF739922.1 | ||||
| BM925434.1 | ||||
| BX461223.2 | ||||
| DA715011.1 | ||||
| DA952579.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038080 | Chr.10:29361590 - 29565510 on Build GRCh38 | Gain |
|
| nsv519798 | Chr.10:29332490 - 29500165 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map