Assay Details
Target Gene Details
Entrez Gene ID: | 10385 |
Gene Name: | butyrophilin subfamily 2 member A2 |
Gene Aliases: |
BT2.2, BTF2, BTN2.2 |
Location: |
Chr.6:26382891-26394874 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BTN2A2 | NM_001197237.1 | 8 | 2030 | NP_001184166.1 |
| NM_001197238.1 | NP_001184167.1 | |||
| NM_001197239.1 | 6 | 1378 | NP_001184168.1 | |
| NM_001197240.1 | 7 | 1879 | NP_001184169.1 | |
| NM_006995.4 | 8 | 2008 | NP_008926.2 | |
| NM_181531.2 | 7 | 1660 | NP_853509.1 | |
| XM_005248797.3 | 8 | 2025 | XP_005248854.1 | |
| XM_005248798.4 | 7 | 1985 | XP_005248855.1 | |
| XM_006714953.3 | 8 | 2277 | XP_006715016.1 | |
| XM_006714955.3 | 8 | 2254 | XP_006715018.1 | |
| XM_011514228.2 | 8 | 2330 | XP_011512530.1 | |
| XM_011514229.2 | 8 | 1839 | XP_011512531.1 | |
| XM_011514231.2 | 6 | 1703 | XP_011512533.1 | |
| XM_017010166.1 | 8 | 2028 | XP_016865655.1 | |
| XM_017010167.1 | 7 | 1930 | XP_016865656.1 | |
| XM_017010168.1 | 8 | 2510 | XP_016865657.1 | |
| XM_017010169.1 | 8 | 2228 | XP_016865658.1 | |
| AK123902.1 | 3 | 2620 | ||
| BC014021.2 | 8 | 2006 | AAH14021.1 | |
| BC017497.1 | AAH17497.1 | |||
| U90550.1 | 8 | 1983 | AAB53428.1 |
Target Gene Details
Entrez Gene ID: | 11118 |
Gene Name: | butyrophilin subfamily 3 member A2 |
Gene Aliases: |
BT3.2, BTF4, BTN3.2, CD277 |
Location: |
Chr.6:26365159-26451697 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BTN3A2 | XM_011514268.2 | XP_011512570.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3608404 | Chr.6:26368701 - 26405919 on Build GRCh38 | Gain |
|
| nsv1023453 | Chr.6:25853602 - 26528022 on Build GRCh38 | Gain |
|
| esv2731730 | Chr.6:26378223 - 26453505 on Build GRCh38 | Deletion |
|
| nsv1030863 | Chr.6:26370778 - 26405762 on Build GRCh38 | Gain+Loss |
|
| esv3608402 | Chr.6:26368480 - 26444267 on Build GRCh38 | Loss |
|
| esv2731728 | Chr.6:26377986 - 26415008 on Build GRCh38 | Deletion |
|
| nsv601184 | Chr.6:26379309 - 26451325 on Build GRCh38 | Gain |
|
| esv3890802 | Chr.6:26369321 - 26442557 on Build GRCh38 | Loss |
|
| nsv970087 | Chr.6:26392104 - 26397135 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map