Assay Details
Target Gene Details
Entrez Gene ID: | 80255 |
Gene Name: | solute carrier family 35 member F5 |
Gene Aliases: |
- |
Location: |
Chr.2:113702677-113756823 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC35F5 | NM_025181.3 | 16 | 2142 | NP_079457.2 |
| NR_104470.1 | ||||
| XM_005263799.3 | 16 | 2527 | XP_005263856.1 | |
| XM_011511921.2 | 17 | 1978 | XP_011510223.1 | |
| XM_011511922.2 | 17 | 2195 | XP_011510224.1 | |
| XM_011511923.2 | 16 | 1880 | XP_011510225.1 | |
| XM_017005027.1 | XP_016860516.1 | |||
| XM_017005028.1 | 15 | 1841 | XP_016860517.1 | |
| AK024666.1 | 11 | 1269 | BAB14954.1 | |
| AY358620.1 | 16 | 1885 | AAQ88983.1 | |
| BC018537.1 | 16 | 2152 | AAH18537.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3592016 | Chr.2:113233328 - 113804636 on Build GRCh38 | Gain |
|
| nsv1001195 | Chr.2:113604426 - 113748753 on Build GRCh38 | Gain |
|
| nsv2882 | Chr.2:113695137 - 113729107 on Build GRCh38 | Insertion |
|
| esv3592028 | Chr.2:113650431 - 113744360 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map