Assay Details
Target Gene Details
Entrez Gene ID: | 6508 |
Gene Name: | solute carrier family 4 member 3 |
Gene Aliases: |
AE3, CAE3/BAE3, SLC2C |
Location: |
Chr.2:219627570-219641980 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC4A3 | NM_001326559.1 | NP_001313488.1 | ||
| NM_005070.3 | NP_005061.2 | |||
| NM_201574.2 | NP_963868.2 | |||
| NR_048551.1 | ||||
| XM_005246789.4 | XP_005246846.1 | |||
| XM_011511665.2 | XP_011509967.1 | |||
| AK226122.1 | ||||
| AK289974.1 | ||||
| AK299109.1 | ||||
| AK314083.1 | ||||
| AY142112.1 | AAN34939.1 | |||
| BC136384.1 | ||||
| BC146656.1 | ||||
| BC171760.1 | ||||
| U05596.1 | AAA50748.1 |
More Information
Set Membership: |
|

Genomic Map