Assay Details
Target Gene Details
Entrez Gene ID: | 59277 |
Gene Name: | netrin 4 |
Gene Aliases: |
PRO3091 |
Location: |
Chr.12:95657807-95790758 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTN4 | NM_021229.3 | 10 | 3483 | NP_067052.2 |
| AF119916.3 | 11 | 3020 | AAF69670.2 | |
| AF278532.1 | 10 | 3485 | AAG30822.1 | |
| AF297711.1 | 10 | 3335 | AAG53651.1 | |
| AK024691.1 | 9 | 2559 | BAB14964.1 | |
| AK026833.1 | 5 | 1829 | ||
| AL137540.1 | 8 | 2351 | CAB70800.3 | |
| AL833767.1 | 9 | 2887 | CAH56243.1 | |
| BC013591.1 | 9 | 2558 | AAH13591.2 |
Target Gene Details
Entrez Gene ID: | 100132594 |
Gene Name: | phosphoglycerate mutase 1 pseudogene 5 |
Gene Aliases: |
- |
Location: |
Chr.12:95649255-95673999 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PGAM1P5 | NR_077225.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv455692 | Chr.12:95627257 - 95680547 on Build GRCh38 | Gain |
|
| nsv1052350 | Chr.12:95528912 - 95686094 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs200886134] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map