Assay Details
Target Gene Details
Entrez Gene ID: | 1478 |
Gene Name: | cleavage stimulation factor subunit 2 |
Gene Aliases: |
CstF-64 |
Location: |
Chr.X:100820359-100841520 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSTF2 | NM_001306206.1 | 12 | 1515 | NP_001293135.1 |
| NM_001306209.1 | 11 | 1404 | NP_001293138.1 | |
| NM_001325.2 | 11 | 1459 | NP_001316.1 | |
| AK095684.1 | ||||
| AK300600.1 | 12 | 1335 | ||
| BC017712.1 | 11 | 1428 | AAH17712.1 | |
| BC033135.1 | 11 | 1348 | AAH33135.1 | |
| BT009778.1 | 11 | 1401 | AAP88780.1 | |
| EU616679.1 | 12 | 1483 | ||
| M85085.1 | 11 | 1423 | AAA35724.1 |
More Information
Set Membership: |
|

Genomic Map