Assay Details
Target Gene Details
Entrez Gene ID: | 10437 |
Gene Name: | IFI30, lysosomal thiol reductase |
Gene Aliases: |
GILT, IFI-30, IP-30, IP30 |
Location: |
Chr.19:18173780-18178124 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFI30 | NM_006332.4 | NP_006323.2 | ||
| AB049659.1 | BAC98466.1 | |||
| AK291551.1 | ||||
| AL558891.3 | ||||
| BE515053.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160592 | Chr.19:18100050 - 18194768 on Build GRCh38 | Deletion |
|
| nsv952420 | Chr.19:18117791 - 18476990 on Build GRCh38 | Deletion |
|
| nsv833774 | Chr.19:18102951 - 18184519 on Build GRCh38 | Loss |
|
| nsv833773 | Chr.19:18042161 - 18192178 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map