Assay Details
Target Gene Details
Entrez Gene ID: | 150468 |
Gene Name: | cytoskeleton associated protein 2 like |
Gene Aliases: |
- |
Location: |
Chr.2:112736349-112764677 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CKAP2L | NM_001304361.1 | 4 | 1330 | NP_001291290.1 |
| NM_152515.4 | 4 | 1389 | NP_689728.3 | |
| NR_130712.1 | 5 | 631 | ||
| XM_011510666.2 | 3 | 1262 | XP_011508968.1 | |
| AK097948.1 | 5 | 571 | BAC05202.1 | |
| AK292530.1 | 4 | 1340 | ||
| AK302875.1 | 4 | 1271 | ||
| AK316241.1 | 3 | 1254 | ||
| AL832036.1 | 4 | 1340 | ||
| BC036217.1 | 4 | 1321 | AAH36217.1 | |
| BF742811.1 |

Genomic Map