Assay Details
Target Gene Details
Entrez Gene ID: | 337972 |
Gene Name: | keratin associated protein 19-5 |
Gene Aliases: |
KAP19.5 |
Location: |
Chr.21:30501872-30502090 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRTAP19-5 | NM_181611.1 | NP_853642.1 | ||
| AB096946.1 | 1 | 211 | BAE46361.1 | |
| BC100836.1 | ||||
| BC100837.2 | ||||
| BC100838.2 | ||||
| BC100839.2 | ||||
| BC113020.1 | ||||
| BC113021.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762104 | Chr.21:30474524 - 30519317 on Build GRCh38 | Gain |
|
| nsv1057467 | Chr.21:30318003 - 30993921 on Build GRCh38 | Gain |
|
| nsv587354 | Chr.21:29978351 - 30620545 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114414951] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map