Assay Details
Target Gene Details
Entrez Gene ID: | 84955 |
Gene Name: | NudC domain containing 1 |
Gene Aliases: |
CML66, OVA66 |
Location: |
Chr.8:109240919-109334205 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDCD1 | NM_001128211.1 | 10 | 3653 | NP_001121683.1 |
| NM_032869.3 | 10 | 3753 | NP_116258.2 | |
| XM_017013910.1 | 11 | 3929 | XP_016869399.1 | |
| XM_017013911.1 | 11 | 3699 | XP_016869400.1 | |
| BC043406.1 | 10 | 3718 | AAH43406.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7281n100 | Chr.8:109157344 - 109290856 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map