Assay Details
Target Gene Details
Entrez Gene ID: | 51474 |
Gene Name: | LIM domain and actin binding 1 |
Gene Aliases: |
EPLIN, SREBP3 |
Location: |
Chr.12:50175780-50283570 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIMA1 | NM_001113546.1 | 11 | 3615 | NP_001107018.1 |
| NM_001113547.1 | 8 | 3496 | NP_001107019.1 | |
| NM_001243775.1 | 5 | 3223 | NP_001230704.1 | |
| NM_016357.4 | 11 | 3612 | NP_057441.1 | |
| XM_011538455.2 | 11 | 3675 | XP_011536757.1 | |
| XM_017019429.1 | 6 | 3125 | XP_016874918.1 | |
| XM_017019430.1 | 11 | 3966 | XP_016874919.1 | |
| AA772616.1 | 1 | 134 | ||
| AF157325.1 | 5 | 3222 | AAF67491.1 | |
| AF198454.1 | 11 | 3539 | AAF23755.1 | |
| AF198455.1 | 8 | 3434 | AAF23756.1 | |
| AK000057.1 | 5 | 2538 | BAA90914.1 | |
| AK000335.1 | 8 | 3136 | BAA91092.1 | |
| AL136911.1 | 11 | 3531 | CAB66845.1 | |
| BC010664.1 | 1 | 1989 | AAH10664.1 | |
| BC110815.1 | 11 | 3549 | AAI10816.1 |
More Information
Set Membership: |
|

Genomic Map