Assay Details
Target Gene Details
Entrez Gene ID: | 7430 |
Gene Name: | ezrin |
Gene Aliases: |
CVIL, CVL, HEL-S-105, VIL2 |
Location: |
Chr.6:158765741-158819424 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EZR | NM_001111077.1 | 14 | 2083 | NP_001104547.1 |
| NM_003379.4 | 13 | 2117 | NP_003370.2 | |
| XM_011536110.1 | 10 | 1494 | XP_011534412.1 | |
| AK129727.1 | 7 | 1039 | ||
| AK296738.1 | ||||
| AK316031.1 | 4 | 840 | ||
| AL162086.1 | 14 | 2035 | CAB82418.1 | |
| BC013903.2 | 13 | 2034 | AAH13903.1 | |
| BC068458.1 | 13 | 2043 | AAH68458.1 | |
| FJ224318.1 | 13 | 2019 | ||
| J05021.1 | 13 | 1978 | AAA61278.1 | |
| X51521.1 | 13 | 2019 | CAA35893.1 |
More Information
Set Membership: |
|

Genomic Map