Assay Details
Target Gene Details
Entrez Gene ID: | 57060 |
Gene Name: | poly(rC) binding protein 4 |
Gene Aliases: |
CBP, LIP4, MCG10 |
Location: |
Chr.3:51957454-51967466 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003453 | Chr.3:51928065 - 52065425 on Build GRCh38 | Gain |
|
| nsv508922 | Chr.3:51940932 - 52009520 on Build GRCh38 | Insertion |
|
| nsv954857 | Chr.3:51932485 - 52056584 on Build GRCh38 | Deletion |
|
| nsv520211 | Chr.3:51738331 - 52132131 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map