Assay Details
Target Gene Details
Entrez Gene ID: | 140807 |
Gene Name: | keratin 72 |
Gene Aliases: |
K6IRS2, K6irs, KRT6, KRT6IRS2 |
Location: |
Chr.12:52585589-52602900 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT72 | NM_001146225.1 | NP_001139697.1 | ||
| NM_001146226.1 | NP_001139698.1 | |||
| NM_080747.2 | NP_542785.1 | |||
| XM_017018833.1 | XP_016874322.1 | |||
| XM_017018834.1 | XP_016874323.1 | |||
| XM_017018835.1 | XP_016874324.1 | |||
| XM_017018836.1 | XP_016874325.1 | |||
| AK093060.1 | BAC04039.1 | |||
| AK293645.1 | ||||
| AK302016.1 | ||||
| AY033495.1 | AAK55108.1 | |||
| BC113686.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428281 | Chr.12:52570654 - 52761443 on Build GRCh38 | Gain |
|
| nsv832415 | Chr.12:52567563 - 52757300 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map