Assay Details
Target Gene Details
Entrez Gene ID: | 114905 |
Gene Name: | C1q and tumor necrosis factor related protein 7 |
Gene Aliases: |
CTRP7, ZACRP7 |
Location: |
Chr.4:15339818-15446169 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1QTNF7 | NM_001135170.1 | 3 | 2997 | NP_001128642.1 |
| NM_001135171.1 | 3 | 2944 | NP_001128643.1 | |
| NM_031911.4 | 3 | 2827 | NP_114117.1 | |
| XM_011513772.1 | 4 | 2867 | XP_011512074.1 | |
| XM_011513773.1 | 4 | 2916 | XP_011512075.1 | |
| BC022187.2 | 3 | 2945 | AAH22187.1 | |
| BC024015.2 | 3 | 2945 | AAH24015.1 | |
| BX647781.1 | 3 | 2997 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759227 | Chr.4:15442993 - 15747897 on Build GRCh38 | Gain+Loss |
|

Genomic Map