Assay Details
Target Gene Details
Entrez Gene ID: | 896 |
Gene Name: | cyclin D3 |
Gene Aliases: |
- |
Location: |
Chr.6:41934933-42048894 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCND3 | NM_001136017.3 | 2 | 473 | NP_001129489.1 |
| NM_001136125.2 | NP_001129597.1 | |||
| NM_001136126.2 | NP_001129598.1 | |||
| NM_001287427.1 | 2 | 976 | NP_001274356.1 | |
| NM_001287434.1 | NP_001274363.1 | |||
| NM_001760.4 | 2 | 476 | NP_001751.1 | |
| XM_011514971.1 | 2 | 471 | XP_011513273.1 | |
| XM_011514972.1 | 2 | 267 | XP_011513274.1 | |
| AB209825.1 | 2 | 267 | BAD93062.1 | |
| AK055519.1 | 2 | 534 | ||
| AK057206.1 | 2 | 267 | ||
| AK096276.1 | 2 | 787 | ||
| AK097856.1 | ||||
| AK299187.1 | ||||
| AK303452.1 | 2 | 255 | ||
| AK315421.1 | 2 | 392 | ||
| BC011616.2 | 2 | 358 | AAH11616.1 | |
| CR542246.1 | 2 | 277 | CAG47042.1 | |
| DA069566.1 | ||||
| DA999482.1 | ||||
| DB001987.1 | ||||
| DB104254.1 | 3 | 449 | ||
| DB110132.1 | ||||
| DC360231.1 | ||||
| M90814.1 | 2 | 377 | AAA51927.1 | |
| M92287.1 | 2 | 442 | AAA52137.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576144 | Chr.6:41934304 - 41947781 on Build GRCh38 | Gain |
|
| nsv830646 | Chr.6:41916050 - 42096473 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map