Assay Details
Target Gene Details
Entrez Gene ID: | 10947 |
Gene Name: | adaptor related protein complex 3 mu 2 subunit |
Gene Aliases: |
AP47B, CLA20, P47B |
Location: |
Chr.8:42152946-42171183 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP3M2 | NM_001134296.1 | 10 | 1514 | NP_001127768.1 |
| NM_006803.3 | 9 | 1455 | NP_006794.1 | |
| XM_017012977.1 | 9 | 1698 | XP_016868466.1 | |
| XM_017012978.1 | 9 | 2480 | XP_016868467.1 | |
| XM_017012979.1 | 8 | 1772 | XP_016868468.1 | |
| AK315228.1 | ||||
| BC056257.1 | 8 | 1147 | AAH56257.1 | |
| BC056398.1 | 9 | 1327 | AAH56398.1 | |
| BX648354.1 | 10 | 1388 | ||
| BX648390.1 | 10 | 1379 | ||
| D38293.1 | 9 | 1313 | BAA07415.1 |
More Information
Set Membership: |
|

Genomic Map