Assay Details
Target Gene Details
Entrez Gene ID: | 51094 |
Gene Name: | adiponectin receptor 1 |
Gene Aliases: |
ACDCR1, CGI-45, CGI45, PAQR1, TESBP1A |
Location: |
Chr.1:202940825-202958572 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADIPOR1 | NM_001290553.1 | 8 | 1504 | NP_001277482.1 |
| NM_001290557.1 | 9 | 1707 | NP_001277486.1 | |
| NM_001290629.1 | 9 | 1593 | NP_001277558.1 | |
| NM_015999.5 | 8 | 1640 | NP_057083.2 | |
| XM_017001407.1 | 9 | 1617 | XP_016856896.1 | |
| AF125179.1 | 8 | 1508 | AAQ13552.1 | |
| AF151803.1 | 8 | 1516 | AAD34040.1 | |
| AK001484.1 | 8 | 1556 | ||
| AK124455.1 | 3 | 1568 | ||
| AK222503.1 | 8 | 1514 | BAD96223.1 | |
| BC001594.2 | 8 | 1533 | AAH01594.1 | |
| BC010743.1 | 8 | 1527 | AAH10743.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114825641] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map