Assay Details
Target Gene Details
Entrez Gene ID: | 79674 |
Gene Name: | ventricular zone expressed PH domain containing 1 |
Gene Aliases: |
MELT, VEPH |
Location: |
Chr.3:157259742-157503626 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VEPH1 | NM_001167911.1 | 13 | 2619 | NP_001161383.1 |
| NM_001167912.1 | 14 | 2754 | NP_001161384.1 | |
| NM_024621.2 | 14 | 2945 | NP_078897.2 | |
| XM_011513134.2 | 13 | 2688 | XP_011511436.1 | |
| XM_011513135.1 | 14 | 2584 | XP_011511437.1 | |
| AB051479.1 | 14 | 2754 | ||
| AK022666.1 | 4 | 622 | BAB14165.1 | |
| AK027625.1 | 14 | 2843 | BAB55243.1 | |
| AL713656.1 | 13 | 2542 | CAD28465.2 | |
| BC042159.1 | 7 | 1404 | AAH42159.1 | |
| BC101660.1 | AAI01661.1 | |||
| BC111017.1 | 13 | 2611 | AAI11018.1 | |
| BC113555.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829766 | Chr.3:157093747 - 157302762 on Build GRCh38 | Gain |
|

Genomic Map