Assay Details
Target Gene Details
Entrez Gene ID: | 285513 |
Gene Name: | GPRIN family member 3 |
Gene Aliases: |
GRIN3 |
Location: |
Chr.4:89237666-89308010 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPRIN3 | NM_198281.2 | 2 | 5640 | NP_938022.2 |
| XM_005262936.3 | 2 | 5880 | XP_005262993.1 | |
| XM_005262937.4 | 2 | 5934 | XP_005262994.1 | |
| XM_017008043.1 | 2 | 7717 | XP_016863532.1 | |
| XM_017008044.1 | 2 | 5438 | XP_016863533.1 | |
| AB095947.2 | ||||
| AK091461.1 | 1 | 1984 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3569577 | Chr.4:89026064 - 89372742 on Build GRCh38 | Loss |
|
| nsv470051 | Chr.4:88723780 - 89721993 on Build GRCh38 | Gain |
|
| nsv830002 | Chr.4:89135081 - 89337714 on Build GRCh38 | Gain |
|
| nsv594806 | Chr.4:88621130 - 90539537 on Build GRCh38 | Gain |
|

Genomic Map