Assay Details
Target Gene Details
Entrez Gene ID: | 127385 |
Gene Name: | olfactory receptor family 10 subfamily J member 5 |
Gene Aliases: |
OR1-28 |
Location: |
Chr.1:159535078-159536007 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR10J5 | NM_001004469.1 | NP_001004469.1 | ||
| BC137025.1 | 1 | 955 | ||
| BC137026.1 | 1 | 955 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3578214 | Chr.1:159515185 - 159615770 on Build GRCh38 | Loss |
|
| nsv1005195 | Chr.1:159482969 - 159940547 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map