Assay Details
Target Gene Details
Entrez Gene ID: | 79841 |
Gene Name: | ATP/GTP binding protein like 2 |
Gene Aliases: |
CCP2 |
Location: |
Chr.11:47659168-47715397 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGBL2 | NM_024783.3 | 19 | 3142 | NP_079059.2 |
| XM_005253138.4 | 17 | 3473 | XP_005253195.1 | |
| XM_005253139.4 | 16 | 3365 | XP_005253196.1 | |
| XM_005253140.4 | 15 | 3239 | XP_005253197.1 | |
| XM_006718328.3 | 15 | 3311 | XP_006718391.1 | |
| XM_011520373.2 | XP_011518675.1 | |||
| XM_011520381.2 | 14 | 3152 | XP_011518683.1 | |
| AK027251.1 | 10 | 1799 | BAB15707.1 | |
| BC028200.1 | 9 | 1347 | AAH28200.1 | |
| BC036234.1 | 19 | 3126 | AAH36234.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469858 | Chr.11:47625963 - 47815620 on Build GRCh38 | Gain |
|
| nsv315 | Chr.11:47630891 - 47660108 on Build GRCh38 | Insertion |
|
| nsv509405 | Chr.11:47571038 - 47663104 on Build GRCh38 | Insertion |
|
| dgv1111n100 | Chr.11:47653918 - 48644472 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map