Assay Details
Target Gene Details
Entrez Gene ID: | 51023 |
Gene Name: | mitochondrial ribosomal protein S18C |
Gene Aliases: |
CGI-134, MRP-S18-1, MRP-S18-c, MRPS18-1, S18mt-c, mrps18-c |
Location: |
Chr.4:83455932-83461723 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS18C | NM_001297767.1 | NP_001284696.1 | ||
| NM_001297769.1 | NP_001284698.1 | |||
| NM_001297770.1 | NP_001284699.1 | |||
| NM_016067.3 | NP_057151.1 | |||
| AF151892.1 | AAD34129.1 | |||
| AK002008.1 | 2 | 1602 | ||
| AK026620.1 | ||||
| BC005186.1 | AAH05186.1 | |||
| BE874586.1 | ||||
| CN262115.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893971 | Chr.4:83096792 - 83493129 on Build GRCh38 | Gain |
|
| esv3601197 | Chr.4:83448066 - 83468701 on Build GRCh38 | Loss |
|
| esv3601196 | Chr.4:83419958 - 83470239 on Build GRCh38 | Loss |
|
| nsv829992 | Chr.4:83244588 - 83458578 on Build GRCh38 | Loss |
|
| nsv829994 | Chr.4:83445146 - 83637163 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map