Assay Details
Target Gene Details
Entrez Gene ID: | 9947 |
Gene Name: | MAGE family member C1 |
Gene Aliases: |
CT7, CT7.1 |
Location: |
Chr.X:141903856-141909401 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEC1 | NM_005462.4 | 4 | 4048 | NP_005453.2 |
| XM_011531418.2 | 4 | 3984 | XP_011529720.1 | |
| AF056334.1 | 1 | 4010 | AAC24227.1 | |
| BQ772869.1 | 1 | 290 |
More Information
Set Membership: |
|

Genomic Map