Assay Details
Target Gene Details
Entrez Gene ID: | 8939 |
Gene Name: | far upstream element binding protein 3 |
Gene Aliases: |
FBP3 |
Location: |
Chr.9:130579567-130638352 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FUBP3 | NM_003934.1 | NP_003925.1 | ||
| XM_005272232.1 | XP_005272289.1 | |||
| XM_006717313.2 | XP_006717376.1 | |||
| XM_011519172.2 | XP_011517474.1 | |||
| XM_011519173.2 | XP_011517475.1 | |||
| XM_011519174.2 | XP_011517476.1 | |||
| XM_017015269.1 | XP_016870758.1 | |||
| XM_017015270.1 | XP_016870759.1 | |||
| XM_017015271.1 | XP_016870760.1 | |||
| XM_017015272.1 | XP_016870761.1 | |||
| XM_017015273.1 | XP_016870762.1 | |||
| AK295155.1 | ||||
| BC001325.1 | AAH01325.1 | |||
| BC007874.2 | AAH07874.1 | |||
| BC137338.1 | ||||
| BC137340.1 | ||||
| U69127.1 | AAC50893.1 |
Target Gene Details
Entrez Gene ID: | 102465515 |
Gene Name: | microRNA 6856 |
Gene Aliases: |
hsa-mir-6856 |
Location: |
Chr.9:130626297-130626363 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR6856 | NR_106915.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054840 | Chr.9:130604380 - 130643160 on Build GRCh38 | Gain |
|
| nsv831735 | Chr.9:130598942 - 130791581 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map