Assay Details
Target Gene Details
Entrez Gene ID: | 5955 |
Gene Name: | reticulocalbin 2 |
Gene Aliases: |
E6BP, ERC-55, ERC55, TCBP49 |
Location: |
Chr.15:76931621-76950260 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RCN2 | NM_001271837.1 | 8 | 1493 | NP_001258766.1 |
| NM_002902.2 | 7 | 1439 | NP_002893.1 | |
| X78669.1 | 7 | 1284 | CAA55343.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760041 | Chr.15:76878209 - 77081106 on Build GRCh38 | Gain |
|
| nsv833058 | Chr.15:76943554 - 77101298 on Build GRCh38 | Loss |
|
| esv2749898 | Chr.15:75764003 - 77933523 on Build GRCh38 | Deletion |
|
| nsv1040852 | Chr.15:76867896 - 77845199 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map