Assay Details
Target Gene Details
Entrez Gene ID: | 10069 |
Gene Name: | RWD domain containing 2B |
Gene Aliases: |
C21orf6, GL011 |
Location: |
Chr.21:29005759-29019371 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RWDD2B | NM_001320724.1 | 5 | 729 | NP_001307653.1 |
| NM_016940.2 | 4 | 624 | NP_058636.1 | |
| AF212232.1 | 4 | 585 | AAK14918.1 | |
| AK000056.1 | 4 | 628 | BAA90913.1 | |
| AK223357.1 | 4 | 635 | BAD97077.1 | |
| BC012546.1 | 4 | 592 | AAH12546.1 | |
| BC017912.1 | 4 | 610 | AAH17912.1 | |
| BI710995.1 | 1 | 196 | ||
| Y19009.1 | 4 | 603 | CAB88085.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893402 | Chr.21:28969569 - 29059824 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map