Assay Details
Target Gene Details
Entrez Gene ID: | 9140 |
Gene Name: | autophagy related 12 |
Gene Aliases: |
APG12, APG12L, FBR93, HAPG12 |
Location: |
Chr.5:115828196-115841851 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATG12 | NM_001277783.1 | 3 | 2097 | NP_001264712.1 |
| NM_004707.3 | 4 | 2234 | NP_004698.3 | |
| NR_033362.1 | 5 | 2351 | ||
| NR_033363.1 | 5 | 2350 | ||
| NR_073603.1 | 6 | 2467 | ||
| BM697565.1 | 1 | 391 | ||
| BX327373.2 | 1 | 724 |
More Information
Set Membership: |
|

Genomic Map